Learn More
Medchemexpress LLC Benzaldehyde, 2,4,5-trimethoxy- | 4460-86-0 | MFCD00003312 | 99.8% | 196.20 g/mol | C10H12O4 | 10 G
SDP
Supplier:  Medchemexpress LLC HY10058010G
Encompass_Preferred
Asaraldehyde (2,4,5-trimethoxybenzaldehyde) is a small-molecule research reagent reported to inhibit cyclooxygenase-2 (COX-2) with an observed IC50 of 100 μg/mL. It is supplied as a high-purity solid for research and analytical applications and is not for human or clinical use.
  • Reported COX-2 inhibitor with IC50 100 μg/mL.
  • High purity (≈99.8%) suitable for analytical work.
  • Molecular formula C10H12O4; molecular weight 196.20 g/mol.
  • Supplied as a 10 G solid package for laboratory use.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-12753