Learn More
Medchemexpress LLC Benzaldehyde, 2,4,6-trimethyl | 487-68-3 | MFCD00003341 | 99.2% | 148.20 g/mol | C10H12O | 500 G

Supplier: Medchemexpress LLC HYW017469500G
Mesitaldehyde (2,4,6-trimethylbenzaldehyde) is an aromatic aldehyde used as a synthetic intermediate and an endogenous metabolite. Supplied as a high-purity reagent, it is suitable for organic synthesis, analytical standards, and research applications.
- High purity: 99.15%
- Molecular formula C10H12O; molecular weight 148.20 g/mol
- Appearance: liquid, colorless to light yellow; density 1.005 g/cm³
- Common applications include organic synthesis and analytical reference material
- Available in laboratory pack sizes such as 100 g, 500 g, and 1 kg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.