Learn More
Medchemexpress LLC Benzaldehyde, 4-fluoro-2-(phenylmethoxy)- | 202857-89-4 | 99.8% | C14H11FO2 | 500 MG

Supplier: Medchemexpress LLC HY76084500MG
2-(Benzyloxy)-4-fluorobenzaldehyde is a drug intermediate for synthesis of various active compounds.
- CAS number: 202857-89-4
- Molecular formula: C14H11FO2
- Molecular weight: 230.24
- Purity: 99.8% (LCMS)
- Appearance: White to off-white solid
- Storage: 4°C, protect from light; in solvent: -80°C, 6 months; -20°C, 1 month (protect from light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.