Learn More
Medchemexpress LLC Benzamide-15N | 31656-62-9 | MFCD00084200 | 50mg
SDP
Supplier:  Medchemexpress LLC HYZ0283S50MG
Encompass_Preferred
Benzamide-15N is a 15N-labeled Benzamide Benzamide inhibits poly(ADP-ribose) polymerase (PARP)[1][2]
Catalog No. 50-002-66265