Learn More
Medchemexpress LLC Benzamide, 2-[[5-bromo-2-[[1-(methylsulfonyl)-4-piperidinyl)amino]-4-pyrimidinyl]amino]-6-fluoro- | 2488892-01-7 | C17H20BrFN6O3S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY151069
Encompass_Preferred
TMX-2039 is a pan-CDK inhibitor for both cell cycle CDKs (CDK1, CDK2, CDK4, CDK5, and CDK6) and transcriptional CDKs (CDK7 and CDK9). It acts as a ligand for target proteins for PROTACs, demonstrating IC50 values against CDK1 (2.6 nM), CDK2 (1.0 nM), CDK4 (52.1 nM), CDK5 (0.5 nM), CDK6 (35.0 nM), CDK7 (32.5 nM), and CDK9 (25 nM).
  • Pan-CDK inhibitor
  • Inhibits cell cycle CDKs
  • Inhibits transcriptional CDKs
  • Ligand for target protein for PROTACs
Catalog No. 50-003-74301