Learn More
Medchemexpress LLC Benzamide, 2-[[(5-bromo-2-thienyl)methylene]amino]- | 304456-62-0 | MFCD00977636 | 98.0% | 309.18 g/mol | C12H9BrN2OS | 25MG

Supplier: Medchemexpress LLC HY12444725MG
BTYNB is a small-molecule inhibitor that blocks IMP1 (IGF2BP1) binding to c-Myc mRNA (reported IC50 = 5 μM). It is supplied for preclinical research and demonstrates selective antiproliferative activity in IMP1-positive cancer cell lines. The compound is provided as a solid with high purity and should be stored protected from light under refrigeration; handle following standard laboratory safety practices.
- Potent IMP1-c-Myc interaction inhibitor (IC50 = 5 μM).
- Selective activity in IMP1-positive cancer cell lines.
- High purity suitable for biochemical and cellular assays.
- Solid form for convenient weighing and storage.
- Storage: protect from light; recommended refrigeration.
- CAS number 304456-62-0 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.