Learn More
Medchemexpress LLC Benzamide, 2-chloro-N-[(S)-phenyl(2S)-2-piperidinylmethyl]-3-(trifluoromethyl)-, hydrochloride | 615571-23-8 | 99.4% | 433.29 g/mol | C20H21Cl2F3N2O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY107155MG
Encompass_Preferred
SSR504734 is a selective, reversible glycine transporter 1 (GlyT1) inhibitor supplied as the hydrochloride salt for research use (CAS 615571-23-8). It shows low-nanomolar potency across human, rat, and mouse GlyT1 and is orally active in preclinical models investigating cognition, anxiety, and depression. The compound is provided as a solid for laboratory studies and should be handled and stored according to standard research-chemical precautions.
  • High purity: 99.4%.
  • Selective, reversible GlyT1 inhibition with low-nanomolar potency.
  • Orally active in animal efficacy models.
  • White to off-white solid physical form.
  • Recommended storage: 4°C, protected from moisture and light.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-40665