Learn More
Medchemexpress LLC Benzamide, 2-((trans-4-hydroxycyclohexyl)amino)-4-(tetrahydro-6,6-dimethyl-4-oxo-indazol-1-yl) | 908112-43-6 | 99.5% | 464.48 g/mol | C23H27F3N4O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1021410MM 1ML
Encompass_Preferred
SNX-2112 is a small-molecule inhibitor of heat shock protein 90 (Hsp90) used in research applications studying Hsp90-mediated signaling and client protein degradation. It is supplied as a 10 mM solution in DMSO (1 mL) and is also available in multiple solid mass formats for experimental flexibility.
  • High purity suitable for research applications, supporting reproducible results.
  • Supplied as a ready-to-use 10 mM solution in DMSO for convenient dosing.
  • Available in multiple solid formats to support different assay types and scales.
  • Molecular weight of 464.48 g/mol for accurate dosing and calculations.
  • Commonly used for cell-based assays and biochemical studies of Hsp90 activity.
Catalog No. 50-002-47117