Learn More
Medchemexpress LLC Benzamide, 3,5-difluoro-N-[4-[(1R)-1-methyl-2-[[(1-methylethyl)sulfonyl)amino]ethyl]phenyl]- | 376594-67-1 | 99.51% | 396.45 | 50 MG

Supplier: Medchemexpress LLC HY10935100MG
LY450108 is a potent AMPA receptor potentiator with potential applications in research related to depression and Parkinson's disease. AMPA receptors are crucial for excitatory neurotransmission and synaptic plasticity in the mammalian central nervous system. Beyond modulating fast synaptic plasticity and memory, AMPA receptor potentiators can influence downstream signaling pathways.
- Target: AMPA receptor potentiator
- Research applications: Potential for depression and Parkinson's disease research
- Form: Solid, white to off-white appearance
- Solubility: Soluble in DMSO (≥ 50 mg/mL), insoluble in H₂O (< 0.1 mg/mL)
- Storage: Powder at -20°C for 3 years, 4°C for 2 years. In solvent, -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.