Learn More
Medchemexpress LLC Benzamide, 3-(aminosulfonyl)-4-chloro-N-(2,3-dihydro-2-methyl-1H-indol-1-yl)- | 26807-65-8 | 99.6% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB02591G
Encompass_Preferred
Indapamide is an orally active sulphonamide diuretic agent that reduces blood pressure by decreasing vascular reactivity and peripheral vascular resistance. It can also reduce left ventricular hypertrophy.
  • Orally active sulphonamide diuretic agent
  • Reduces blood pressure by decreasing vascular reactivity and peripheral vascular resistance
  • Can reduce left ventricular hypertrophy
  • High purity of 99.6%
  • Appears as a white to off-white solid
  • Soluble in DMSO (≥ 100 mg/mL)
  • Demonstrates biological activity in vitro, including reducing insulin secretory response and increasing osteoblast proliferation
  • Shown to lower blood pressure in spontaneously hypertensive rats in vivo
Catalog No. 50-004-05379