Learn More
Medchemexpress LLC Benzamide, 4-[[1-[[2-chloro-5-(trifluoromethyl)phenyl]methyl]-4-piperidinyl]methoxy]-5-cyclopropyl | 1788071-27-1 | 98.9% | C25H27ClF4N2O4S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13187010MG
Encompass_Preferred
GX-201 is a selective NaV1.7 inhibitor for research use that potently inhibits human NaV1.7 (IC50 <3.2 nM). The compound produces analgesia in mouse pain models (formalin and CFA) after oral dosing and is supplied as a high-purity solid with recommended cold storage for stability.
  • Selective NaV1.7 inhibition (IC50 <3.2 nM).
  • Demonstrated analgesic activity in formalin and CFA mouse models.
  • High purity (98.89%).
  • Solid form with recommended cold storage for stability.
  • Suitable for in vitro and in vivo preclinical research.
Catalog No. 50-002-61024