Learn More
Medchemexpress LLC Benzamide, 4-(4-cyanophenoxy)- | 1042780-52-8 | 98.0% | 238.24 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1487535MG
Encompass_Preferred
PARP10-IN-2 is a potent mono-ADP-ribosyltransferase PARP10 inhibitor with an IC50 of 3.64 μM for human PARP10. It also shows inhibition on PARP2 and PARP15 with IC50s of 27 μM and 11 μM for human PARP2 and human PARP15, respectively.
  • Potent mono-ADP-ribosyltransferase PARP10 inhibitor
  • Inhibits PARP2 and PARP15
  • Cytoprotective activity against human HeLa cells
Catalog No. 50-003-62650