Learn More
Medchemexpress LLC Benzamide | 55-21-0 | MFCD00007968 | 200mg
SDP
Supplier:  Medchemexpress LLC HYZ0283200MG
Encompass_Preferred
Benzamide (Benzenecarboxamide) is a potent poly(ADP-ribose) polymerase (PARP) inhibitor Benzamide has protective activity against both glutamate- and methamphetamine (METH)-induced neurotoxicity in vitro Benzamide can attenuate the METH-induced dopamine depletions and exhibits neuroprotective activity in mice also has no acute effect on striatal dopamine metabolism and does not reduce body temperature[1]
Catalog No. 50-002-66224