Learn More
Medchemexpress LLC Benzamide, n-[(3,4-dichlorophenyl)methyl]-5-(6-fluoro-2-methyl-3-pyridinyl) trifluoroacetate | 2749300-35-2 | 98.0% | 612.40 g·mol-1 | C27H23Cl2F4N5O3 | 5 MG

Supplier: Medchemexpress LLC HY111753A5MG
WDR5-IN-4 TFA is a small-molecule inhibitor of the WIN site of WD repeat-containing protein 5 (WDR5), supplied as the trifluoroacetate salt for research use. Provided in a 5 mg pack, it features high reported purity and defined molecular properties suitable for biochemical and cellular studies of WDR5-dependent epigenetic regulation.
- High reported purity (98.0%).
- Supplied as the trifluoroacetate (TFA) salt.
- Molecular weight 612.40 g·mol-1.
- Molecular formula C27H23Cl2F4N5O3.
- Suitable for biochemical and cellular assays.
- Provided in a 5 mg pack for small-scale research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.