Learn More
Medchemexpress LLC Benzamide, N-[4-[(4-ethyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-4-methyl-3-[[[5-(2-thienyl... | 1186206-79-0 | 99.01% | 607.69 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY18007100MG
Encompass_Preferred
ALW-II-41-27 is an Eph family tyrosine kinase inhibitor with an IC50 of 11 nM for Eph2. This compound is suitable for research applications targeting tyrosine kinases.
  • Inhibits Ba/F3 cells transformed with Tel fusions of EphA3, Kit, Fms, KDR, FLT1, FGR, Src, Lyn, Bmx, and Bcr-Abl.
  • Exhibits cross-reactivity with Bcr-Abl.
  • Inhibits a broad range of kinases including b-raf, CSF1R, DDR1, DDR2, EphA2, EphA5, EphA8, EphB1, EphB2, EphB3, Frk, Kit, Lck, p38α, p38β, PDGFRα, PDGFRβ, and Raf1.
Catalog No. 50-003-86814