Learn More
Medchemexpress LLC Benzamidine hydrochloride | 1670-14-0 | MFCD00013025 | 99.9% | 156.61 g/mol | C7H9ClN2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW018781100MG
Encompass_Preferred
Benzamidine hydrochloride is the hydrochloride salt of benzamidine, a reversible competitive inhibitor of trypsin-like serine proteases. It is used in biochemical and enzymology research to inhibit proteases such as trypsin, tryptase, urokinase (uPA), Factor Xa, thrombin, and tPA, and is supplied in solid and solution formats for assay use.
  • Reversible competitive inhibitor of trypsin-like serine proteases.
  • Useful for enzymology, protease inhibition assays, and biochemical studies.
  • Reported Ki values provide comparative potency against common serine proteases.
  • Available as solid and as DMSO solution formats for assay convenience.
  • High purity (>99.9%) suitable for research applications.
  • Store solid at 4°C under nitrogen; solutions long-term at -80°C.
Catalog No. 50-003-00712