Learn More
Medchemexpress LLC Benzamidine hydrochloride hydrate | 206752-36-5 | MFCD00066285 | 99.6% | 156.61 g/mol | C7H8N2·HCl·xH2O | 10 G
SDP
Supplier:  Medchemexpress LLC HYW08793710G
Encompass_Preferred
Benzamidine hydrochloride hydrate is a reversible competitive inhibitor of trypsin-like serine proteases, supplied as a crystalline hydrated hydrochloride. It is used in biochemical and cell-based assays to inhibit trypsin, tryptase, uPA, Factor Xa, thrombin, and tPA, and has been reported to reduce thymidine incorporation in fibroblasts and to decrease tryptase-stimulated collagen synthesis.
  • Reversible competitive inhibitor of trypsin-like serine proteases.
  • Reported inhibitory activity against tryptase, trypsin, uPA, Factor Xa, thrombin, and tPA.
  • High reported purity (about 99.6%).
  • Provided as a hydrate; molecular weight depends on water content.
  • Available in small lab pack sizes suitable for research use (e.g., 5 g, 10 g).
Catalog No. 50-002-08405