Learn More
Cayman Chemical BenzamIlhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1431310 MG
Encompass_Preferred
An ENaC blocker that inhibits sodium transport from (IC50 = 4 nM) and binds to (Kd = 5 nM) bovine kidney cortex membrane vesicles; an NCX blocker that inhibits the cytosolic calcium response to extracellular sodium depletion in mouse podocytes (IC50 ~ 100 nM); an inhibitor of TRPP3 (IC50 = 1.1 μM)
Catalog No. 50-295-3842