Learn More
Medchemexpress LLC Benzarone | 1477-19-6 | >98% | 266.29 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW011711100MG
Encompass_Preferred
Benzarone is identified as an oral inhibitor of human urate transporter 1 (hURAT1), possessing an IC50 value of 2.8 μM. It also functions as an uncoupler of oxidative phosphorylation.
  • Oral inhibitor of human urate transporter 1 (hURAT1).
  • Acts as an uncoupler of oxidative phosphorylation.
  • Can be utilized to reduce serum uric acid levels.
  • Useful for research in vascular diseases.
Catalog No. 50-004-53677