Learn More
Medchemexpress LLC Benzarone | 1477-19-6 | 99.67% | 266.29 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01171110MM/1ML
Encompass_Preferred
Benzarone is an oral inhibitor of human urate transporter 1 (hURAT1) with an IC50 value of 2.8 μM, and it also acts as an uncoupler of oxidative phosphorylation. Benzarone can cause liver damage and promote cell apoptosis and necrosis. It can be used to lower serum uric acid levels and for research in vascular diseases.
  • Oral inhibitor of human urate transporter 1 (hURAT1)
  • IC50 value of 2.8 μM for hURAT1 in oocyte
  • Acts as an uncoupler of oxidative phosphorylation
  • Can cause liver damage and promote cell apoptosis and necrosis
  • Used to lower serum uric acid levels
  • Used for research in vascular diseases
Catalog No. 50-004-52870