Learn More
Medchemexpress LLC Benzenamine, 3-methyl-4-(4-piperidinyl)- | 1260864-79-6 | MFCD17486190 | 98.0% | 190.29 | C12H18N2 | 10 MG

Supplier: Medchemexpress LLC HYI087510MG
Ceritinib impurity 5 is an analytical reference impurity of the ALK inhibitor ceritinib, provided as a research-use-only laboratory chemical for analytical and synthetic applications. It is characterized and supplied with COA and SDS documentation for traceability and safe handling.
- Analytical impurity standard for ceritinib.
- Purity 98.0% by certificate of analysis.
- Molecular weight 190.29 and formula C12H18N2.
- Light blue to blue solid physical appearance.
- Intended for laboratory research and manufacture of substances.
- Hazard statements include harmful if swallowed, causes skin and eye irritation, and may cause respiratory irritation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.