Learn More
Medchemexpress LLC Benzenamine, homopolymer | 25233-30-1 | MFCD00241446 | 98.6% | C8H11N | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01776750G
Encompass_Preferred
Polyaniline is a conductive polymer supplied as a reference compound for materials science and antimicrobial research. It is reported with a high purity and is provided with batch-specific quality documentation and safety information to support laboratory use and reproducibility.
  • Conducting polymer for materials and electronics research.
  • Useful precursor or component in formulations for antibacterial agents.
  • Reported purity of 98.6% for analytical confidence.
  • Supplied with certificate of analysis for batch verification.
  • Includes safety data sheet for handling and storage guidance.
  • Available in laboratory-scale packaging for R&D use.
Catalog No. 50-003-00906