Learn More
Medchemexpress LLC Benzene, 1-(1,1-dimethylethyl)-3-ethoxy- | 133073-81-1 | 95.0% | C12H18O | 10 G

Supplier: Medchemexpress LLC HY7598110G
1-tert-Butyl-3-ethoxybenzene is a drug intermediate used for the synthesis of various active compounds. It is for research use only and not intended for human use. The appearance is liquid, with a color ranging from light yellow to light brown.
- Used for synthesis of various active compounds
- For research use only
- Appearance: liquid, light yellow to light brown
- Pure form storage: -20°C for 3 years or 4°C for 2 years
- In solvent storage: -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.