Learn More
Medchemexpress LLC Benzene, 1,2-difluoro-4-nitro- | 369-34-6 | MFCD00007198 | 99.9% | 159.091 g/mol | C6H3F2NO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW015472
Encompass_Preferred
3,4-Difluoronitrobenzene (CAS 369-34-6) is a nitro-substituted difluorobenzene used as a building block and drug intermediate in organic synthesis, notably for the preparation of oxazolidinone-class antibiotics. It is supplied as a colorless to light yellow liquid and is characterized for use in research and synthetic applications.
  • High purity suitable for synthetic applications.
  • Useful drug intermediate for oxazolidinone antibiotic synthesis.
  • Liquid form simplifies handling and measurement.
  • Well-characterized: molecular formula C6H3F2NO2 and molecular weight 159.091 g/mol.
  • Certificate of analysis reports assay 99.86% by GC.
Catalog No. 50-003-03088