Learn More
Medchemexpress LLC Benzene-1,3,5-tricarboxylic acid | 554-95-0 | 99.8% | C9H6O6 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY7898510MM/1ML
Encompass_Preferred
Benzene-1,3,5-tricarboxylic acid (Trimesic acid) is a scaffold. It enhances the binding ability of bicyclic peptides to proteins and improves the mechanical stability of hydrogels.
  • Enhances the binding ability of bicyclic peptides to proteins.
  • Improves the mechanical stability of hydrogels.
Catalog No. 50-004-34509