Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzene, 2,4-dimethoxy-1-methyl- | 38064-90-3 | MFCD00010749 | ≥97.0% | 152.19 g/mol | C9H12O2 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW3244245G

Encompass_Preferred

2,4-Dimethoxytoluene is an aromatic biochemical reagent used in laboratory research for synthesis and analytical applications. It is typically a colorless to pale-yellow liquid supplied in small quantities for bench-scale work and is intended for research use only.

  • Chemical name: Benzene, 2,4-dimethoxy-1-methyl-.
  • CAS number: 38064-90-3.
  • Molecular formula C9H12O2; molecular weight 152.19 g/mol.
  • Purity ≥97.0% by assay.
  • Available in small pack sizes such as 5 G for bench-scale use.
  • Store at room temperature (solid); in solvent, refrigerate or freeze for long-term storage.

Catalog No. 50-002-80797


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.