Learn More
Medchemexpress LLC Benzene, 2-bromo-4-fluoro-1-methoxy- | 452-08-4 | 205.026 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW001094
Encompass_Preferred
2-Bromo-4-fluoroanisole, also known as Benzene, 2-bromo-4-fluoro-1-methoxy-, is a chemical compound with a molecular formula of C7H6BrFO and a molecular weight of 205.026. This compound typically appears as a colorless to light yellow liquid and is supplied with a purity of 99.65% (GC). It is suitable for various laboratory research applications.
  • High purity for reliable results
  • Long shelf life when stored correctly
  • Liquid form for easy handling
  • Suitable for laboratory research
Catalog No. 50-004-50014