Learn More
Medchemexpress LLC Benzeneacetamide, 4-fluoro-.alpha.-(4-fluorophenyl)-.alpha.-phenyl- | 289656-45-7 | 99.9% | C20H15F2NO | 1 ML

Supplier: Medchemexpress LLC HY5069410MM/1ML
Senicapoc (ICA-17043) is a potent and selective Gardos channel (Ca2+-activated K+ channel; KCa3.1) blocker with an IC50 of 11 nM. It blocks Ca2+-induced rubidium flux from human red blood cells with an IC50 value of 11 nM and inhibits red blood cell dehydration with an IC50 of 30 nM. This product is intended for research use only.
- Potent and selective Gardos channel blocker
- Blocks calcium-induced rubidium flux from human red blood cells
- Inhibits red blood cell dehydration
- Molecular formula: C20H15F2NO
- CAS number: 289656-45-7
- Purity: 99.9%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.