Learn More
Medchemexpress LLC Benzeneacetamide, N-[(1R)-4-[(aminoiminomethyl)amino]-1-[[1R)-1-(4-hydroxyphenyl)ethyl]amino] | 00-00-0 | MFCD04112988 | 99.7% | 533.62 g/mol | C29H35N5O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY101704A50MG
Encompass_Preferred
Y1 receptor antagonist 1 formic is a small-molecule research compound that selectively inhibits the neuropeptide Y (NPY) Y1 receptor. It is provided as a purified formic salt and is intended for in vitro pharmacology and receptor-signaling studies, with analytical characterization for identity and purity.
  • Selective Y1 receptor antagonism suitable for receptor pharmacology studies.
  • High purity as confirmed by HPLC for consistent experimental results.
  • Supplied as a formic salt to aid solubility in polar solvents.
  • Characterized by molecular formula C29H35N5O5 and molecular weight 533.62 g/mol.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-47779