Learn More
Medchemexpress LLC Benzeneacetamide, N-[3-(aminosulfonyl)-4-(3-chlorophenoxy)phenyl]-2,6-dichloro- | 2055601-42-6 | 99.9% | 485.77 | 5 MG

Supplier: Medchemexpress LLC HY1606455MG
P2X4 antagonist-1 (Compound 24) is an antagonist for P2X4 with an IC50 of 15 nM. It is for research use only and not sold to patients. This compound demonstrates antagonist activity at human P2X4 receptor transfected in HEK293 cells. The product is suitable for various applications including compound screening and cellular effect assays.
- Molecular weight: 485.77
- Formula: C20H15Cl3N2O4S
- CAS number: 2055601-42-6
- Appearance: Solid, white to off-white
- Target: P2X4 receptor, with an IC50 of 15 nM
- Applications: Compound screening libraries (Immunology/Inflammation, Membrane Transporters/Ion Channels, Neurotransmitter Receptors, Bioactive Compound Libraries)
- Applied in cellular effect assays with HEK293 cell lines to assess inhibition of Bz-ATP-induced calcium influx
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.