Learn More
Medchemexpress LLC Benzeneacetic acid, α,3,4-trihydroxy- | 775-01-9 | 98.0% | 184.147 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11347410MM/1ML
Encompass_Preferred
3,4-Dihydroxymandelic acid is a metabolite of norepinephrine. It is classified as an Endogenous Metabolite and is relevant to Metabolic Disease research. The standard version of 3,4-Dihydroxymandelic acid is intended for research and analytical applications.
  • Endogenous Metabolite relevant to Metabolic Disease research
  • Suitable for research and analytical applications
Catalog No. 50-004-36810