Learn More
Medchemexpress LLC Benzeneacetic acid, 3-methyl- | 621-36-3 | MFCD00004340 | 99.9% | 150.18 g/mol | C9H10O2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0535075G
Encompass_Preferred
m-Tolylacetic acid (3-methylbenzeneacetic acid) is a solid analytical standard and chemical intermediate used in organic synthesis and analytical applications. It has molecular formula C9H10O2, molecular weight 150.18 g/mol, and CAS number 621-36-3. The material is supplied with high purity suitable for research use.
  • High purity suitable for analytical and synthetic applications.
  • Solid form convenient for storage and weighing.
  • Commonly used as a building block in organic synthesis.
  • Available in small-scale 5 g packaging for lab use.
Catalog No. 50-002-10944