Learn More
Medchemexpress LLC Benzeneacetic acid cyclopentyl ester (CHR-2845) | 914382-60-8 | 99.7% | 495.61 g/mol | C28H37N3O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10640910MM 1ML
Encompass_Preferred
Tefinostat is a monocyte/macrophage-targeted histone deacetylase (HDAC) inhibitor prodrug used in preclinical research. It is converted intracellularly to the active acid metabolite and is supplied in research-ready formats for biochemical and cellular studies.
  • Monocyte/macrophage-targeted HDAC inhibition suitable for immuno-oncology research.
  • Prodrug that is cleaved intracellularly to the active acid metabolite.
  • Available as a 10 mM solution in DMSO or as solid material for flexibility in use.
  • High purity for reproducible experimental results.
  • Supplied with a certificate of analysis for batch-specific quality data.
Catalog No. 50-002-40926