Learn More
Medchemexpress LLC Benzeneacetic acid, α-hydroxy-, phenylmethyl ester, (αR)- | 97415-09-3 | MFCD00674031 | 98.0% | 242.27 | C15H14O3 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0206555G
Encompass_Preferred
(R)-Benzyl mandelate is the R-enantiomer of benzyl mandelate (D-(-)-mandelic acid benzyl ester). It is a chiral synthetic intermediate used in pharmaceutical and organic synthesis for building blocks and preparation of optically active compounds. The material is supplied as a white to off-white solid and is characterized by NMR-confirmed purity and defined storage conditions.
  • R-enantiomer suitable for enantioselective synthesis.
  • High purity (NMR 98.0%).
  • White to off-white solid physical form.
  • Molecular formula C15H14O3 and molecular weight 242.27.
  • Stable storage: room temperature up to 3 years; in solvent -80°C (2 years) or -20°C (1 year).
Catalog No. 50-003-00936