Learn More
Medchemexpress LLC Benzeneacetic acid, α-hydroxy-, phenylmethyl ester (αR) | 97415-09-3 | MFCD00674031 | 98.0% | 242.27 g·mol⁻¹ | C15H14O3 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02065510G
Encompass_Preferred
(R)-Benzyl mandelate (D-(-)-mandelic acid benzyl ester) is the R-enantiomer of benzyl mandelate used as a chiral synthetic intermediate in pharmaceutical and fine chemical synthesis. It has CAS 97415-09-3, molecular formula C15H14O3, and molecular weight 242.27 g·mol⁻¹. The reagent is supplied at high reported purity for research and synthetic applications.
  • High chemical purity (98.0%) for reliable synthetic outcomes.
  • Defined R-enantiomer for enantioselective synthesis.
  • Useful chiral building block for pharmaceutical intermediates.
  • Reported molecular weight and formula support accurate stoichiometry.
  • Available in small-scale quantities suitable for research use.
Catalog No. 50-003-00740