Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-d3, sodium salt (1:1) | 1219805-09-0 | 99.2% | C₁₃H₁₄D₃NaO₂ | 1 MG
SDP

Supplier:  Medchemexpress LLC HY78131CS1MG

Encompass_Preferred

This Ibuprofen-d3 sodium product is a white to off-white solid. It exhibits high purity and isotopic enrichment, complying with specified analytical standards.

  • Appearance: White to off-white solid
  • Purity (HPLC): 99.21%
  • Isotopic enrichment: 99.9%
  • Recommended storage: 4°C, sealed, away from moisture
  • Recommended storage in solvent: -80°C for up to 6 months; -20°C for up to 1 month, sealed and away from moisture

Catalog No. 50-004-34557


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.