Learn More
Medchemexpress LLC Benzeneacetonitrile, 4-[(4-methoxyphenyl)methoxy]- | 175135-47-4 | 98.1% | 253.30 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY18771200MG
Encompass_Preferred
O4I1 is a potent Oct3/4 inducer that enhances Oct3/4 gene expression, increases Oct3/4 protein levels, and promotes Oct3/4 mediated transcriptional activation. It activates Oct3/4 at both transcriptional and translational levels in various human cell lines and is sufficient to enhance Oct3/4 overexpression by 2-fold and 3-fold after 48 and 72 hours of treatment, respectively.
  • Potent Oct3/4 inducer.
  • Enhances Oct3/4 gene expression.
  • Increases Oct3/4 protein levels.
  • Promotes Oct3/4 mediated transcriptional activation.
  • Activates Oct3/4 at transcriptional and translational levels in diverse human cell lines.
  • Can enhance Oct3/4 overexpression 2-fold and 3-fold after 48 and 72 hours of treatment in untransfected HEK293 cells.
Catalog No. 50-003-88214