Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzeneacetonitrile, 4-chloro- | 140-53-4 | MFCD00001918 | 99.8% | 151.59 g/mol | C8H6ClN | 1000 G
SDP

Supplier:  Medchemexpress LLC HYW0173901000G

Encompass_Preferred

4-Chlorobenzyl cyanide (4-chlorophenylacetonitrile) is an aromatic nitrile used as a synthetic intermediate and biochemical reagent for research. It is supplied at high analytical purity and accompanied by batch QC data suitable for laboratory synthesis and analytical applications.

  • High purity for research (99.84% GC).
  • Characterized by 1H NMR and water content analysis (~0.12% by KF).
  • Available in multiple pack sizes, including 1,000 g.
  • Physical form: solid below ~25°C and liquid above ~28°C; colorless to off-white.
  • Recommended storage prolongs stability (4°C short term, -20°C long term).

Catalog No. 50-003-01376


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.