Learn More
Medchemexpress LLC Benzenebutanaminium, 4-chloro-N,N-diethyl-N-heptyl-, 4-methylbenzenesulfonate (1:1) | 92953-10-1 | 99.6% | 510.17 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY333505MG
Encompass_Preferred
Clofilium tosylate is a potassium channel blocker that induces apoptosis of human promyelocytic leukemia (HL-60) cells via Bcl-2-insensitive activation of caspase-3. It is also an antiarrhythmic agent.
  • Potassium channel blocker.
  • Induces apoptosis in HL-60 cells.
  • Acts via Bcl-2-insensitive activation of caspase-3.
  • Antiarrhythmic agent.
Catalog No. 50-004-04471