Learn More
Medchemexpress LLC Benzeneethanamine, 3,4-dimethoxy-N-methyl-N-[3-[4-[[1-methyl-2-(1-methylethyl)-1H-indol-3-yl]sulfonyl] | 121345-64-0 | 99.8% | 564.74 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13690950MG
Encompass_Preferred
SR33805 is a potent Ca2+ channel antagonist with EC50s of 4.1 nM and 33 nM in depolarized and polarized conditions, respectively. It selectively blocks L-type but not T-type Ca2+ channels, making it suitable for research on acute or chronic failing hearts.
  • Potent Ca2+ channel antagonist.
  • Selectively blocks L-type Ca2+ channels.
  • Useful for research on acute or chronic failing hearts.
  • Inhibits growth factor-induced proliferation of smooth muscle cells (SMC) in vitro in a dose-dependent manner.
  • Restores myocardial infarction (MI)-altered cell shortening without affecting Ca2+ transient amplitude in vitro.
  • Improves end-systolic strain and fractional shortening of MI hearts in rats in vivo.
  • Significantly reduces intimal hyperplasia in pigs in vivo.
Catalog No. 50-003-52084