Learn More
Medchemexpress LLC Benzeneethanol, 4-hydroxy- | 501-94-0 | 138.16 | 250 MG

Supplier: Medchemexpress LLC 250MGHYN0474
Tyrosol is a derivative of phenethyl alcohol. It attenuates pro-inflammatory cytokines from cultured astrocytes and NF-κB activation, demonstrating anti-oxidative and anti-inflammatory effects.
- Attenuates pro-inflammatory cytokines from cultured astrocytes
- Attenuates NF-κB activation
- Exhibits anti-oxidative and anti-inflammatory effects
- Targets NF-κB and endogenous metabolite
- Research areas include inflammation/immunology
- Disease research fields include anti-aging
- Structurally classified as monophenols, phenols
- Sourced from microorganisms, plants, Canarium album (Lour.) Rauesch., Burseraceae
- Appears as a white to off-white solid
- Purity (HPLC) of 99.93%
- Water (KF) content is 0.07%
- Residue on ignition is 0.01%
- Consistent with structure by ¹H NMR spectrum
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.