Learn More
Medchemexpress LLC Benzeneethanol, β-[[5-(1H-indazol-5-yl)-3-pyridinyl)amino]-, (βR)- | 1613638-82-6 | 99.4% | 330.38 | 10 MG

Supplier: Medchemexpress LLC HY11146510MM/1ML
CDK8-IN-4 is a potent CDK8 inhibitor, with an IC50 of 0.2 nM, derived from patent WO2014090692A1, compound example 16. It is presented as a white to light yellow solid.
- Inhibits CDK8 with an IC50 of 0.2 nM
- Molecular formula: C20H18N4O
- Molecular weight: 330.38
- Target: CDK
- Pathway: Cell Cycle/DNA Damage
- Purity: 99.39% by HPLC
- Recommended storage: 4°C, sealed, away from moisture and light; in solvent: -80°C (6 months) or -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.