Learn More
Medchemexpress LLC Benzenemethanaminium, N,N,N-triethyl-, chloride | 56-37-1 | MFCD00011824 | ≥98.0% | 227.77 | C13H22ClN | 250g
SDP
Supplier:  Medchemexpress LLC HYY0667250G
Encompass_Preferred
Benzyltriethylammonium chloride is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Benzyltriethylammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-69115