Learn More
Medchemexpress LLC Benzenemethanol, 4-hydroxy-3-methoxy- | 498-00-0 | 154.16 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN206710MM/1ML
Encompass_Preferred
Vanillyl alcohol is an orally active phenolic alcohol that reduces ROS generation, suppresses Bax, and increases Bcl-2. It exhibits anti-angiogenic, anti-inflammatory, anti-nociceptive, and neuroprotective effects, and is also used as a flavoring agent in foods and beverages.
  • Increases viability of MPP+-induced MN9D dopaminergic cells
  • Inhibits acetic acid-induced vascular permeability
  • Reduces exudate volume and polymorphonuclear leukocytes in carrageenan-induced air pouches
Catalog No. 50-004-14961