Learn More
Medchemexpress LLC Benzenemethanol, 4-hydroxy-3-methoxy- | 498-00-0 | 154.16 | 100 G
SDP
Supplier:  Medchemexpress LLC HYN2067100G
Encompass_Preferred
Vanillyl alcohol is an orally active phenolic alcohol known for its anti-angiogenic, anti-inflammatory, anti-nociceptive, and neuroprotective properties. It also serves as a flavoring agent in various foods and beverages.
  • Reduces ROS generation
  • Suppresses Bax and increases Bcl-2
  • Exhibits anti-angiogenic effects
  • Exhibits anti-inflammatory effects
  • Exhibits anti-nociceptive effects
  • Exhibits neuroprotective effects
  • Increases viability of MPP+-induced MN9D dopaminergic cells in vitro
  • Inhibits vascular permeability and writhing response in animal models
  • Reduces inflammation markers in animal models
Catalog No. 50-004-15715