Learn More
Medchemexpress LLC Benzenepropanoic acid, 4-methoxy-, ethyl ester | 22767-72-2 | 250 MG

Supplier: Medchemexpress LLC 250MGHYN3869
Ethyl-p-methoxyhydrocinnamate is a compound identified as Benzenepropanoic acid, 4-methoxy-, ethyl ester. This product has a purity of 97.64% by HPLC and appears as a colorless to light yellow liquid. It is suitable for various laboratory applications requiring high purity.
- High purity: 97.64% (HPLC)
- Appearance: Colorless to light yellow liquid
- Molecular formula: C12H16O3
- Molecular weight: 208.26
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.