Learn More
Medchemexpress LLC Benzenepropanol | 122-97-4 | 99.8% | 136.19 | 250 G

Supplier: Medchemexpress LLC 250GHY41304
Benzenepropanol is a biochemical reagent suitable for life science related research. It is intended for research use only and not for patient use. This product is provided as a liquid with a density of 1.001 g/cm3, typically colorless to light yellow.
- High purity of 99.76%
- Available in various quantities for research needs
- Can be stored at -20°C for 3 years in pure form
- Soluble in DMSO for in vitro applications
- Multiple in vivo dissolution methods available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.