Learn More
Medchemexpress LLC Benzenesulfinic acid, sodium salt (1:1) | 873-55-2 | 99.97% | 164.16 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW013598
Encompass_Preferred
Sodium benzenesulfinate is a reagent employed in organic synthesis for the creation of other compounds, such as sulfones. It participates in rhodium-catalyzed desulfinative coupling reactions with aldehydes, resulting in the production of ketones.
  • Reagent for organic synthesis
  • Used to create sulfones
  • Undergoes rhodium-catalyzed desulfinative coupling with aldehydes to yield ketones
Catalog No. 50-004-51468