Learn More
Medchemexpress LLC Benzenesulfonamide, 2,4-difluoro-N-[2-methoxy-5-[4-[3-[4-(methylsulfonyl)-1-piperazinyl]-1- | 1676893-24-5 | 100.0% | 627.68 g·mol⁻¹ | C29H27F2N5O5S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100470100MG
Encompass_Preferred
NSC781406 is a small-molecule dual inhibitor of phosphoinositide 3-kinase (PI3K) and mechanistic target of rapamycin (mTOR) for research use. It exhibits low-nanomolar enzymatic potency and cellular antiproliferative activity, and has shown antitumor efficacy in preclinical xenograft models.
  • Potent PI3Kα inhibition (IC50 2 nM).
  • Dual PI3K/mTOR activity (mTOR IC50 5.4 nM).
  • Broad cellular antiproliferative activity across cancer cell lines.
  • High chemical purity (99.97%).
  • Solid form for standard laboratory handling.
  • Supplied as a 100 mg research package.
Catalog No. 50-002-14745