Learn More
Medchemexpress LLC Benzenesulfonamide, N-[(1R,2R)-2-amino-1,2-diphenylethyl]-4-methyl- | 144222-34-4 | 366.48 | 10 G

Supplier: Medchemexpress LLC HYW00287810G
TsDPEN (Compound 2) is a biochemical reagent. It can be applied to copper catalyzed asymmetric Kinugasa reactions using its N,N,N imide chiral ligands. This product is a white to off-white solid with a molecular weight of 366.48.
- Biochemical reagent for research applications
- Applied in copper catalyzed asymmetric Kinugasa reactions
- Purity of 99.49%
- Store at 4°C, protected from light, under nitrogen
- In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light, under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.